Learn More
Thermo Scientific™ Smart-Vue™ Receivers
Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.
Brand:  Thermo Scientific™ SV102-504-LSB
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11894663
  • 889.00€ / Each
Estimated Shipment
24-02-2021
Add to basket
Description
Description
Compatible with:
Refrigerators, Freezers, LN2 Storage, CO2 Incubators, Clean Rooms, Storage Areas, Ovens
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Specifications
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Universal power supply
Receiver, 868MHz
RS232 25/500mW Receiver w/universal power supply
868MHz