Learn More
Honeywell™ SKELETON NIT 3 Gloves
Honeywell™ SKELETON NIT 3 Gloves feature a shock absorber pad on top of the hand to protect metacarpian and fingers.
Brand:  Honeywell™ 2332275-10
Additional Details : Weight : 0.16200kg
 View more versions of this product
Product Code. 15816785
  • 16.68€ / 1 pair
Estimated Shipment: 08-11-2021
Add to basket
Description
Description
  • Knitted glove in Spectra™
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The Spectra™ yarn offers very good resistance to cut
  • Black nitrile foam coating on palm
  • Nitrile coating provides good protection against oil and grease
  • The foam coating makes the glove more flexible than standard nitrile
  • Rubber pad on top of the hand and fingers
  • Helps to protect metacarpian and fingers from shocks
Specifications
Specifications
Blue, White
Nitrile
25 cm
13 gauge
10
EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards