Learn More
Thermo Scientific™ SkanIt™ Software for Microplate Readers
Microplate reader control and data handling is simple and intuitive with the SkanIt Software, providing excellent usability and flexibility even for the most challenging microplate assays. SkanIt software helps you to optimize your research with an easy-to-read visual workflow, effortless data analysis, and flexible exporting capabilities.
Brand:  Thermo Scientific™ N18600
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15823547
  • 3665.00€ / Each
Estimated Shipment: 20-09-2021
Add to basket
Description
Description
With SkanIt Software, you have full control over the instrument settings for all your Thermo Scientific™ microplate readers.  SkanIt™ software supports optimal use of the instrument features, offers visual workflow setup and effortless data reduction and exporting. Operation is simple and straightforward even for new users who only require basic reading capabilities. SkanIt Software Upgrade Package is specifically designed for Thermo Scientific Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers to enable the use of SkanIt software and USB connection between instrument and PC. SkanIt Software is available in two editions: a research edition for scientists working in life science research and a drug discovery edition with features for compliancy with FDA 21 CFR Part 11. 
  • Multi-instrument software platform compatible with all Thermo Scientific microplate readers
  • Open license software for unlimited installation to multiple computers
  • Research Edition for life sciences; Drug Discovery Edition with features for compliance with FDA 21 CFR Part 11
  • User-interface languages available: English, French, German, Spanish, Portuguese, Italian, Russian, Japanese, and Chinese
  • Optional separate user accounts for controlling access
  • Robotic automation interface
  • Searchable SkanIt Cloud Library with ready-to-use protocols including calculations
  • Measurement sessions are easy to setup and navigate
  • Intuitive Virtual Pipetting Tool assists in defining the plate layout
  • Capable of endpoint, kinetic, spectral, bottom-reading multipoint, and kinetic-spectral measurements
  • Single- or multiple-wavelength measurements and even multi-technology measurements
  • Results data show in real-time during the measurement
  • Merge data from multiple measurements into one kinetic data set
  • Transfer data to an Excel file with a single click
  • Manually or automatically export data to .xlsx, .pdf, .xml, and .txt file formats
  • Automatically load measurement data into a predefined Excel template file
  • SkanIt Software Upgrade Package is also compatible with the discontinued Fluoroskan Ascent, Fluoroskan Ascent FL, and Luminoskan Ascent microplate readers.
    • Note: the upgrade package is compatible with Ascent instruments that are equipped with internal software version 2.6.
Specifications
Specifications
SkanIt Software upgrade package, drug discovery edition
Microplate Reader
Fluoroskan, Fluoroskan FL, Luminoskan, Fluoroskan Ascent, Fluoroskan Ascent FL, Luminoskan Ascent
SkanIt Software Upgrade Package