missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Tamper Evident Self-Standing Transport Tubes

For all applications requiring tamper evident seal for utmost security and sample integrity

Brand:  Simport™ Scientific T552-30ATTP

Additional Details : Weight : 5.90000kg

 View more versions of this product

Product Code. 15808712

  • 92.22€ / Pack of 500


Estimated Shipment: 14-10-2021
Add to basket

Description

Description

  • For secure short and long term storage
  • Provide strength and clarity
  • Non-toxic, non sterile polypropylene made etched or non graduated tubes
  • Exhibit unique design features
  • With external threads for providing smooth and uniform inner surface
  • Closure offers unique frangible band design
  • Combination of tamper evident and resistant closure system
  • For clinical trials and expensive reagents in diagnostic kits
  • Caps sold separately

CAPS SOLD SEPARATELY

These unique transport tubes incorporate a seal that needs to be broken in order to open it, which leaves visual indication that the vial has been opened.

Specifications

Specifications

1.01 oz.
25.3 mm
Self-Standing Transport Tube
30 mL
Polypropylene
111 mm
Non-sterile
Polyethylene
Yes
Autoclavable
Conical
Storage and transportation of biological materials
500/Cs.
Videos
SDS
Documents
Certificates