missing translation for 'onlineSavingsMsg'
Learn More

Silver Bromide, 99%, Honeywell™ Fluka™

Catalog Number 15622250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
7785-23-1
AgBr
187.772
MFCD00003398
ADZWSOLPGZMUMY-UHFFFAOYSA-M
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
66199
bromosilver
Br[Ag]
This item is not returnable. View return policy
7785-23-1
AgBr
187.772
MFCD00003398
ADZWSOLPGZMUMY-UHFFFAOYSA-M
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
66199
bromosilver
Br[Ag]

0.99

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 432°C
Physical Form Solid
Quantity 25 g
UN Number NONH for all modes of transport
Formula Weight 187.77g/mol
Percent Purity 99%
Packaging Poly Bottle
Density 6.473g/mL at 25°C
Chemical Name or Material Silver Bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'