Learn More
Sampling Systems™ Silicone Seals for Mini-Drums
These seals when used with toggle clamps will make mini-drum containers leak-proof. Choose the right sized seal for your mini-drum.
Brand: Sampling Systems™ SIL-2-FDA
Additional Details : Weight : 0.04000kg
Description
- O-ring design
- Made from FDA-acceptable grade silicone
- Suitable for use with both 304 and 316L stainless steel mini-drums
- Wide range of sizes available
Specifications
Leak-proof seals | |
Seal, O-ring | |
Silicone | |
2L mini-drum |