missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Silicone Seals for Mini-Drums

These seals when used with toggle clamps will make mini-drum containers leak-proof. Choose the right sized seal for your mini-drum.

Brand:  Sampling Systems™ SIL-15-FDA

Additional Details : Weight : 0.04000kg

 View more versions of this product

Product Code. 15809731

Quantity Price
1 31.84€ / Each
10 26.89€ / Each
Estimated Shipment: 03-11-2021

Add to basket


Description

Description

  • O-ring design
  • Made from FDA-acceptable grade silicone
  • Suitable for use with both 304 and 316L stainless steel mini-drums
  • Wide range of sizes available
Specifications

Specifications

Leak-proof seals
15L mini-drum
Silicone
O-ring Seal
Videos
SDS
Documents
Certificates