Learn More
Uvex™ Shirt Women 8999-Chili
Catalog Number 17206581
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
The uvex suXXeed collection gets everything spot on: well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove. Can be worn in industrial and metal processing settings as well as for leisure
  • T-shirt
  • Color: navy
  • Material: 60% lyocell (TENCEL™), 40% polyester, ca. 180 g/m²
  • T-Shirt made from a high-functional and lightweight knitted fabric with a timeless design.
  • Sporty fit, round neck with contrasting piping, very elastic and lightweight fabric for excellent wearing comfort, easy-care
  • The TENCEL™-fibre reduces the growth of micro-organisms and the development of odour on a natural way
  • Oeko-Tex Standard 100
  • Sizes: XS - 3XL
  • Care instructions: 60°C gently washable, chlorinate impossible, tumble dryer: 1 point, ironing: 1 point, dry cleaning: impossible
Size 3X-Large
Gender Women's
Color Orange
Disposable No
Thickness 160 g/m²