missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Shirt with Short Sleeve 7484-Black

Catalog Number 17206061
Click to view available options
Size:
3X-Large/4X-Large
5X-Large/6X-Large
Medium/Large
Small
X-Large/2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove.

  • Shirt men
  • Color: Black
  • Material: 68% Dryarn, 28% Tencel, 4% Elasthan, 200 gsm
  • Seamless underwear without annoying seams
  • Moisture-regulating
  • Blend with natural fibers
  • Integrated functional zones
  • Maximum comfort
  • OEKO-TEX™ Standard 100
  • Sizes: S - 5XL/6XL
  • Care: washable at 30 ° C, not chlorine, not bleaching, not drying not ironing, not cleaning
Size Small
Color Black
Thickness (Metric) 200 g/m²
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'