missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminohydantoin, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against Aminohydantoin.
Brand: Abnova PAB8220.100uL
Additional Details : Weight : 0.02000kg
Specifications
| Aminohydantoin | |
| Unconjugated | |
| Enzyme Immunoassay (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Liquid | |
| Aminohydantoin conjugated with BSA. | |
| RUO | |
| Primary | |
| Not Provided |
| ELISA | |
| Sheep polyclonal antibody raised against Aminohydantoin. | |
| In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide) | |
| Sheep | |
| 100 uL | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Polyclonal |
For Research Use Only