Learn More
Honeywell™ SHARPFLEX Liner Gloves
Honeywell™ SHARPFLEX Liner Gloves for handling heavy parts with high cut risks in a dry environment.
Brand:  Honeywell™ 2232522-11
Additional Details : Weight : 0.06200kg
 View more versions of this product
Product Code. 15816735
  • 5.88€ / 1 pair
Estimated Shipment: 08-11-2021
Add to basket
Description
Description
  • Knitted glove in composite and Kevlar yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The Kelvar yarn assure a resistance to the contact heat and cut risks
  • Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
  • Being Ambidexterous increases the lifetime of the glove
Specifications
Specifications
Green, Yellow
Polyamide
25 cm
Ambidextrous
13 gauge, Dupont™ Kevlar™ aramid fibers and composite fiber
11
EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards, EN 407-04 Thermical hazards