missing translation for 'onlineSavingsMsg'
Learn More

Sesamol, 98%

Product Code. p-7080774
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11428673

Brand: Thermo Scientific Alfa Aesar A10453.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 533-31-3 | C7H6O3 | 138.122 g/mol

Sesamol has good antioxidant activity. Sesamol has potential protective effects against free radicals as well as some antifungal activity. Sesamol can also be used as an intermediate in the preparation of antidepressants such as Paroxetine (P205750). Sesamol has been found to be an antioxidant that may prevent the spoilage of oils. It also may prevent the spoilage of oils by acting as an antifungal. It is also used in ayurvedic Medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sesamol has good antioxidant activity. Sesamol has potential protective effects against free radicals as well as some antifungal activity. Sesamol can also be used as an intermediate in the preparation of antidepressants such as Paroxetine (P205750). Sesamol has been found to be an antioxidant that may prevent the spoilage of oils. It also may prevent the spoilage of oils by acting as an antifungal. It is also used in ayurvedic Medicine.

Solubility
Soluble in methanol, water (partly), and dichloromethane (25 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-31-3
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00005827
InChI Key LUSZGTFNYDARNI-UHFFFAOYSA-N
Synonym sesamol, 3,4-methylenedioxy phenol, 5-hydroxy-1,3-benzodioxole, 3,4-methylenedioxyphenol, 2h-1,3-benzodioxol-5-ol, 3,4-methylendioxyphenol, methylene ether of oxyhydroquinone, phenol, 3,4-methylenedioxy, 5-benzodioxolol, unii-94iea0nv89
PubChem CID 68289
ChEBI CHEBI:9126
IUPAC Name 1,3-benzodioxol-5-ol
SMILES C1OC2=C(O1)C=C(C=C2)O

Specifications

Melting Point 61°C to 65°C
Boiling Point 113°C to 116°C (2 mmHg)
Quantity 10 g
Beilstein 127405
Sensitivity Light sensitive
Solubility Information Soluble in methanol,water (partly),and dichloromethane (25mg/ml).
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material Sesamol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.