Learn More
Hamilton™ Septa, Long Life GC Autosampler
103.00€
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10491963
|
Hamilton™
75832 |
11.5 mm |
103.00€
Pack of 25 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
10268132
|
Hamilton™
75831 |
9 mm |
103.00€
Pack of 25 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||