missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 334867
GREENER_CHOICE

Product Code. 18738096
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
This item is not returnable. View return policy

Product Code. 18738096

Brand: Tocris Bioscience™ 1960/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective non-peptide OX1 antagonist

SB 334867 is a selective non-peptide orexin OX1 receptor antagonist. pKb values are 7.2 and < 5 for inhibition of intracellular Ca2+ release in CHO cells expressing human OX1 and OX2 receptors respectively. Blocks orexin-A induced grooming and feeding following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Methyl-6-benzoxazolyl)-N'-1,5-naphthyridin-4-yl urea
CAS 792173-99-0
Assay Percent Range >99%
Quantity 1 mg
Solubility Soluble to 100mM with gentle warming in DMSO
Molecular Formula C17H13N5O2
Formula Weight Observed MW: 319.32

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.