Learn More
Ark-Plas Products™ Safety Spiral Wrap
Formulated from polyethylene. Ark-Plas Products™ Safety Spiral Wrap is perfect for protecting against abrasions for wires, hoses, and tubings.
Brand:  Ark-Plas Products™ WXX69-TE6
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11898541
  • 161.27€ / Each
Estimated Shipment
07-04-2021
Add to basket
Description
Description
  • Weather resistant polyethylene
  • Abrasion protection for wires
Specifications
Specifications
Fluorescent Yellow
1 mm
0.070 mm
Polyethylene
75 mm