missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N α-Dimethylbenzylamine, 99+%, Thermo Scientific Chemicals

Catalog Number p-4812523
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19131-99-8
C9H13N
135.21
RCSSHZGQHHEHPZ-QMMMGPOBSA-N
s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
2060073
(1S)-N-methyl-1-phenylethanamine
CC(C1=CC=CC=C1)NC
This item is not returnable. View return policy
19131-99-8
C9H13N
135.21
RCSSHZGQHHEHPZ-QMMMGPOBSA-N
s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
2060073
(1S)-N-methyl-1-phenylethanamine
CC(C1=CC=CC=C1)NC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9240g/mL
Boiling Point 74.0°C to 76.0°C (11.0 mmHg)
Flash Point 61°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NHCH3
Quantity 1 g
Specific Gravity 0.924
Solubility Information Solubility in water: insoluble
Formula Weight 135.21
Percent Purity 99+%
Chemical Name or Material (S)-(-)-N α-Dimethylbenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'