Learn More
Abeba™ Rubber 9310 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
Brand: Abeba™ 9310/44
Additional Details : Weight : 0.95000kg
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- Black
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather insole
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li | |
Leather | |
Slip-resistant | |
9310 |
Black | |
44 | |
Rubber |