missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RPS14 Recombinant Protein
229.00€ - 508.00€
Specifications
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
---|---|
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Molecular Weight (g/mol) | 42.35 |
pH Range | 8 |
Preparation Method | In vitro wheat germ expression system |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16190362
|
Abnova™
H00006208-P01.2UG |
2μg |
229.00€
2µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16154715
|
Abnova™
H00006208-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16164715
|
Abnova™
H00006208-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blot | |
42.35 | |
In vitro wheat germ expression system | |
Wheat Germ (in vitro) | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Human | |
Solution |