missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ROBO1 Pre-design Chimera RNAi
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants (NM_001145845.1,NM_002941.3,NM_133631.3)
POA
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
For Use With (Application) | RNAi Knockdown |
Description
- Gene name: ROBO1
- Gene Alias: DUTT1, FLJ21882, MGC131599, MGC133277, SAX3
- Entrez GeneID: 6091
- Gene Description: Roundabout, axon guidance receptor, homolog 1
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
RNAi Knockdown |
For research use only