Learn More
Rnase-free Water, non DEPCAbnova™
RNase-free Water is suitable to experiment and research of RNA.
Brand:  Abnova R0013.500mL
Additional Details : Weight : 0.02000kg
Quantity 500 mL
 View more versions of this product
Código de producto. 16022048
  • 110.16€ / 500mL
Estimated Shipment: 20-01-2022
Añadir a la cesta
Descripción
Descripción
  • The RNase-free Water, non DEPC was prepared from highly purified water by ultrafiltration without DEPC treatment.
  • The product was confirmed to be free from RNase and DNase.
  • Apperance is clear liquid.
Especificaciones
Identificadores de productos químicos
Especificaciones
500 mL
Videos
SDS
Documentos
Certificados
Ofertas especiales
Ofertas especiales