Learn More
Thermo Scientific™ Revco™ Enzyme Freezer
Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired.
POA
Specifications
Type | Enzyme Freezer |
---|---|
Temperature Range | -20°C |
Door Style | Solid |
No. of Doors | 1 |
Cabinet Material | Scratch-resistant Painted |
Description
Choose Thermo Scientific™ Revco™ Enzyme Freezers for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. Our manual defrost −20°C upright freezers feature microprocessor control system, gravity air circulation, industrial-quality cabinet construction and extra-strength refrigeration compressors designed for industrial, clinical and scientific use.
Key Features- Operating temperature: -20°C
- Preset temperature setpoint
- Upright 23.3 cu. ft. (659L) capacity
- Gravity convection air system for maximum stability without blowers or fans
- Manual defrost
Microprocessor Control System:
- Icon-based controls and displays
- Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature
- Large, easy-to-read digital display of temperature within 0.1°C
- Audible/visual warnings for overtemperature, undertemperature, or power failure
- Key-operated OFF/ON/ALARM ON master switch for setpoint security
- Push-button alarm test
- Remote alarm contacts
- Alarm silence, ringback, and auto reset
- Battery backup system
Construction:
- Scratch-resistant, painted cold-rolled steel interior and exterior finishes
- Solid door with full-length handle and keyed lock—self-closes against closed-cell peripheral gasket
- High-density, CFC-free urethane foam insulation protects cabinet from ambient temperature fluctuations and minimizes operating costs
- Hermetically sealed heavy-duty, industrial-grade compressor
- Nine epoxy-coated, open-wire shelves, adjustable in 1in. (2.5cm) increments
- Adjustable leveling feet
Ordering Information:
Alarm systems, temperature chart recorders, and power monitors available separately. Contact your Customer Service or Sales Representative for details.
Includes:
45 high-impact plastic enzyme storage bins for easily accessible inventory. Bin overall dimensions (L × W × H): 25.25 × 4.25 × 3in. (64 × 11 × 8cm); labels not provided.
Specifications
Enzyme Freezer | |
Solid | |
Scratch-resistant Painted | |
Epoxy Coated Steel | |
Upright | |
Warranty varies by country, please contact us for details | |
29 in. | |
58 in. | |
94.5 cm | |
Heavy-Duty, Industrial-Grade, Hermetically Sealed |
-20°C | |
1 | |
Urethane Foam | |
9 | |
Microprocessor | |
Manual | |
79.2 in. | |
37.2 in. | |
Alarm, Recorder | |
CFC-free |