Learn More
RETSCH Economy Clamping Device for AS 200 Sieve Shaker
Provides reliable, quick and convenient clamping of sieves in dry operation.
Brand:  RETSCH 32.662.0003
Additional Details : Weight : 3.60000kg
Product Code. 10014500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Threaded rods and clamping nuts
  • Stainless-steel cover
Specifications
Specifications
Test Sieves,100 to 203 mm dia.
Economy Clamping Device
Reliable, quick, convenient clamping of sieves in dry operation
AS 200 sieve shaker