Learn More
Eppendorf™ Research™ Plus (EU-IVD), Multi-Channel, Variable
The ultra-light Eppendorf Research plus mechanical pipette not only meets the highest needs in precision and accuracy, it also provides you with increased flexibility and ultimate ergonomics.
880.00€ - 935.00€
Specifications
| Adjustable Tip Spacing | No |
|---|---|
| Autoclavable | Yes |
| Finger Style | Ergonomic |
| Model | Eppendorf Research™ plus (EU-IVD) |
| Product Type | Manual Pipette |
| Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | ||||
| 16202391 |
Eppendorf™
3122000078 |
20 µL to 1 µL | 16 Channels |
880.00€ Each |
|
16222391 |
Eppendorf™
3122000094 |
100 µL to 5 µL | 16 Channels |
880.00€ Each |
|
16212391 |
Eppendorf™
3122000086 |
20 µL to 1 µL | 24 Channels |
935.00€ Each |
|
16232391 |
Eppendorf™
3122000108 |
100 µL to 5 µL | 24 Channels |
935.00€ Each |
|
Description
- Air-displacement pipette for accurate pipetting of aqueous solutions
- Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
- Spring loaded tip cone (available for all pipettes up to 1 mL) for minimal tip attachment forces helps to reduce stress
- Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
- Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
- Autoclave the entire pipette or only the lower part according to your needs to ensure decontamination
- With the channel display, you can always use the pipette in the same position.
- Increase your flexibility and remove individual channels as necessary
Specifications
| No | |
| Ergonomic | |
| Manual Pipette |
| Yes | |
| Eppendorf Research™ plus (EU-IVD) |
The Research™ plus pipette is designed and constructed for low-contamination transfer of liquids, in particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order to allow the in-vitro diagnostic medical device to be used as intended. This pipette is an in vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998. It is intended exclusively for indoor use and for operation by qualified staff.