Learn More
Thermo Scientific™ Replacement Parts for Model 960
Brand:  Thermo Scientific™ 96014
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10059150
  • 162.18€ / Each
Estimated Shipment: 19-01-2022
Add to basket
Description
Description
Automatic Stirrer Probe can also be used with Orion Star Meters.
Specifications
Specifications
Tubing Kit
Tube and connector for working solution bottle to dispenser, tube and connectors for dispenser to dispenser probe