Learn More
Cole-Parmer™ Replacement Ionic Strength adjustors
Ensure consistent and accurate readings from your ISE with these ionic strength adjustors
Brand:  Cole-Parmer™ 27503-51
Additional Details : Weight : 0.01000kg
Product Code. 11709719
  • 50.05€ / Each
Estimated Shipment: 12-06-2024
Description
  • Replacement ionic strength adjustor
  • 5 M NaNO3
  • Replacement ionic strength adjustors come in 475mL bottles (except model 27503-58, which comes in a one-gallon bottle)
Specifications
Specifications
Reference Electrode: 5 M NaNO3
Replacement Ionic Strength Adjustors
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • SODIUM NITRATE ADJUSTOR 5M
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : sodium nitrate