Learn More
Binder™ Reinforced Rack
Binder™ Reinforced Rack for use with a range of Binder™ incubators, growth chambers, freezers, drying and heating chambers and climate chambers.
Brand:  Binder™ 8012-0828
Additional Details : Weight : 5.50000kg
 View more versions of this product
Product Code. 15621926
  • 290.00€ / Each
please call +351214253350
Add to basket
Description
Description
  • Manufactured from stainless steel
  • With one set of securing elements (4 pieces)
  • Maximum load of 70kg
Specifications
Specifications
Stainless steel
Rack
1 set of 4 fasteners
KT 115, KT 170, KT 53
Max. 70kg
Videos