Learn More
Abeba™ Reflexor™ 6900 Shoes
Antistatic shoe includes PU-sole with high slip resistance.
Brand:  Abeba™ 6900/41
Additional Details : Weight : 0.85000kg
 View more versions of this product
Product Code. 15622408
  • 116.40€ / 1 pair
Estimated Shipment
24-06-2019
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 SRC
  • Smooth leather white
  • Slip resistance PU outsole sole
  • 2 replaceable Reflexor™ insoles (article no. 3565/ 3567)
  • Antistatic
  • Lining microfiber, respiration absorbing
  • Instep area with lacing
  • Hand-sewn seam
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRC
Microfiber
41
Polyurethane
White
Leather
Slip resistant
6900
Videos