missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Proliferating cell nuclear antigen Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Proliferating cell nuclear antigen, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Accession Number | NP_009645.1 |
---|---|
Gene ID (Entrez) | 852385 |
Name | S. cerevisiae Proliferating cell nuclear antigen Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023540
|
enQuireBio™
QP6471-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16063540
|
enQuireBio™
QP6471-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16033540
|
enQuireBio™
QP6471-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16053540
|
enQuireBio™
QP6471-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16073540
|
enQuireBio™
QP6471-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16043540
|
enQuireBio™
QP6471-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_009645.1 | |
S. cerevisiae Proliferating cell nuclear antigen Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
852385 | |
Research Use Only | |
POL30 | |
Recombinant Protein | |
Yeast | |
MLEAKFEEAS LFKRIIDGFK DCVQLVNFQC KEDGIIAQAV DDSRVLLVSL EIGVEAFQEY RCDHPVTLGM DLTSLSKILR CGNNTDTLTL IADNTPDSII LLFEDTKKDR IAEYSLKLMD IDADFLKIEE LQYDSTLSLP SSEFSKIVRD LSQLSDSINI MITKETIKFV ADGDIGSGSV IIKPFVDMEH PETSIKLEMD QPVDLTFGAK YLLDIIKGSS LSDRVGIRLS SEAPALFQFD LKSGFLQFFL APKFNDEE | |
Greater than 90% as determined by SDS-PAGE. |