missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-lyase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Cystathionine beta-lyase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7238-ec-1mg
Detalles adicionales : Weight : 0.01000kg
Especificaciones
NP_011331.3 | |
S. cerevisiae Cystathionine beta-lyase Protein | |
Research Use Only | |
STR3 | |
Recombinant Protein | |
E. coli | |
MPIKRLDTVV VNTGSQNDQH SASVPPVYLS TTFKVDLNNE DAQNYDYSRS GNPTRSVLQH QIGKLYRVPQ ENVLAVSSGM TALDVILRGL VLLNGTDNHT PTIIAGDDLY GGTQRLLNFF KQQSHAVSVH VDTSDFEKFK TVFQSLDKVD CVLLESPTNP LCKVVDIPRI LRFVKCISPD TTVVVDNTMM SGLNCNPLQL NPGCDVVYES ATKYLNGHHD LMGGVIISKT PEIASKLYFV INSTGAGLSP MDSWLLVRGL KTLGVRLYQQ QRNAMILAHW LENSCGFKPT RTNKATKTRF VGLRSNPDFK LHKSFNNGPG AVLSFETGSF EHSKRLVSSK KLSIWAVTVS FGCVNSLLSM PCKMSHASID PELRKERDFP EDLVRLCCGI ENIVDLKKDL LAAMVDADII EVRENGKYLF NKLNKNLAVN TTIDDLHKPL SIYEEFYNQD LIRKDSELNI KSSKL | |
Greater than 90% as determined by SDS-PAGE. |
852691 | |
1 mg | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |