missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Prostatic Acid Phosphatase / ACPP Protein

A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Prostatic Acid Phosphatase / ACPP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6412-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023130

Quantity Price
1 1037.95€ / 100µg
4 976.40€ / 100µg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

WP_000426914.1
S. aureus Prostatic Acid Phosphatase / ACPP Protein
100 μg
Not determined.
Full Length
S. aureus
His
Tris-based buffer, 50% glycerol
31213993
Greater than 90% as determined by SDS-PAGE.
Research Use Only
acpP
Recombinant Protein
Yeast
MENFDKVKDI IVDRLGVDAD KVTEDASFKD DLGADSLDIA ELVMELEDEF GTEIPDEEAE KINTVGDAVK FINSLEK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers