missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabF Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7667-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030081

Quantity Price
1 3211.16€ / 1mg
3 3021.38€ / 1mg
Estimated Shipment: 19-01-2022

Add to basket


Description

Description

Specifications

Specifications

WP_000081240.1
S. aureus fabF Protein
1 mg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
31213734
Greater than 90% as determined by SDS-PAGE.
Research Use Only
fabF
Recombinant Protein
E. coli
MSQNKRVVIT GMGALSPIGN DVKTTWENAL KGVNGIDKIT RIDTEPYSVH LAGELKNFNI EDHIDKKEAR RMDRFTQYAI VAAREAVKDA QLDINENTAD RIGVWIGSGI GGMETFEIAH KQLMDKGPRR VSPFFVPMLI PDMATGQVSI DLGAKGPNGA TVTACATGTN SIGEAFKIVQ RGDADAMITG GTEAPITHMA IAGFSASRAL STNDDIETAC RPFQEGRDGF VMGEGAGILV IESLESAQAR GANIYAEIVG YGTTGDAYHI TAPAPEGEGG SRAMQAAMDD AGIEPKDVQY LNAHGTSTPV GDLNEVKAIK NTFGEAAKHL KVSSTKSMTG HLLGATGGIE AIFSALSIKD SKVAPTIHAV TPDPECDLDI VPNEAQDLDI TYAMSNSLGF GGHNAVLVFK KFEA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers