missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Alpha-hemolysin Protein

A DNA sequence encoding the Staphylococcus aureus (strain NCTC 8325) Alpha-hemolysin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7634-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019920

  • 3405.00€ / 1mg
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

WP_000857483.1
S. aureus Alpha-hemolysin Protein
Research Use Only
hly
Recombinant Protein
E. coli
ADSDINIKTG TTDIGSNTTV KTGDLVTYDK ENGMHKKVFY SFIDDKNHNK KLLVIRTKGT IAGQYRVYSE EGANKSGLAW PSAFKVQLQL PDNEVAQISD YYPRNSIDTK EYMSTLTYGF NGNVTGDDTG KIGGLIGANV SIGHTLKYVQ PDFKTILESP TDKKVGWKVI FNNMVNQNWG PYDRDSWNPV YGNQLFMKTR NGSMKAADNF LDPNKASSLL SSGFSPDFAT VITMDRKASK QQTNIDVIYE RVRDDYQLHW TSTNWKGTNT KDKWIDRSSE RYKIDWEKEE MTN
Greater than 90% as determined by SDS-PAGE.
3920722
1 mg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers