missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat SerpinF1 / PEDF Protein
A DNA sequence encoding the Rattus norvegicus (Rat) SerpinF1 / PEDF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6675-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_808788.1 | |
SerpinF1 / PEDF Protein | |
Research Use Only | |
Serpinf1 | |
Recombinant Protein | |
E. coli | |
NVPDSSQDSP APDSTGEPVV EEDDPFFKAP VNKLAAAVSN FGYDLYRLRS GAVSTGNILL SPLSVATALS ALSLGAEQRT ESVIHRALYY DLINNPDIHS TYKELLASVT APEKNFKSAS RIVFERKLRV KSSFVAPLEK SYGTRPRILT GNPRIDLQEI NNWVQAQMKG KIARSTREMP SALSILLLGV AYFKGQWATK FDSRKTTLQD FHLDEDRTVR VPMMSDPKAI LRYGLDSDLN CKIAQLPLTG SMSIIFFLPL TVTQNLTMIE ESLTSEFVHD IDRELKTIQA VLTVPKLKLS YEGDVTNSLQ DMKLQSLFES PDFSKITGKP VKLTQVEHRA AFEWNEEGAG TSSNPDLQPV RLTFPLDYHL NRPFIFVLRD TDTGALLFIG RILDPSST | |
Greater than 90% as determined by SDS-PAGE. |
287526 | |
100 μg | |
Not determined. | |
Full Length | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |