missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Nitric oxide synthase, endothelial Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Nitric oxide synthase, endothelial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8865-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016891

  • 725.49€ / 100µg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_068610.1
Nitric oxide synthase, endothelial Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
24600
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Nos3
Recombinant Protein
E. coli
ATILYGSETG RAQSYAQQLG RLFRKAFDPR VLCMDEYDVV SLEHEALVLV VTSTFGNGDP PENGESFAAA LMEMSGPYNS SPRPEQHKSY KIRFNSVSCS DPLVSSWRRK RKESSNTDSA GALGTLRFCV FGLGSRAYPH FCAFARAVDT RLEELGGERL LQLGQGDELC GQ
Videos
SDS
Documents
Certificates
Special Offers

Special Offers