missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Lutropin-choriogonadotropic hormone receptor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Lutropin-choriogonadotropic hormone receptor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6306-ma-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037110.1 | |
Lutropin-choriogonadotropic hormone receptor Protein | |
Research Use Only | |
Lhcgr | |
Recombinant Protein | |
Mammal | |
RELSGSRCPE PCDCAPDGAL RCPGPRAGLA RLSLTYLPVK VIPSQAFRGL NEVVKIEISQ SDSLERIEAN AFDNLLNLSE LLIQNTKNLL YIEPGAFTNL PRLKYLSICN TGIRTLPDVT KISSSEFNFI LEICDNLHIT TIPGNAFQGM NNESVTLKLY GNGFEEVQSH AFNGTTLISL ELKENIYLEK MHSGAFQGAT GPSILDISST KLQALPSHGL ESIQTLIALS SYSLKTLPSK EKFTSLLVAT LTYPSHCCAF RNLPKKEQNF SFSIFENFSK QCESTVRKAD NETLYSAIFE ENELSGWDYD YGFCSPKTLQ CAPEPDAFNP CEDIMG | |
Greater than 90% as determined by SDS-PAGE. |
25477 | |
20 μg | |
Not determined. | |
Full Length | |
Rat | |
Flag | |
Tris-based buffer, 50% glycerol |