missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Lutropin-choriogonadotropic hormone receptor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Lutropin-choriogonadotropic hormone receptor, was expressed in the hosts and tags indicated.
605.00€ - 1690.00€
Specifications
Accession Number | NP_037110.1 |
---|---|
Gene ID (Entrez) | 25477 |
Name | Lutropin-choriogonadotropic hormone receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032350
|
enQuireBio™
QP6306-MA-20UG |
20 μg |
605.00€
20µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16022350
|
enQuireBio™
QP6306-MA-100UG |
100 μg |
1690.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16042350
|
enQuireBio™
QP6306-MA-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_037110.1 | |
Lutropin-choriogonadotropic hormone receptor Protein | |
Not determined. | |
Full Length | |
Rat | |
Flag | |
Tris-based buffer, 50% glycerol |
25477 | |
Research Use Only | |
Lhcgr | |
Recombinant Protein | |
Mammal | |
RELSGSRCPE PCDCAPDGAL RCPGPRAGLA RLSLTYLPVK VIPSQAFRGL NEVVKIEISQ SDSLERIEAN AFDNLLNLSE LLIQNTKNLL YIEPGAFTNL PRLKYLSICN TGIRTLPDVT KISSSEFNFI LEICDNLHIT TIPGNAFQGM NNESVTLKLY GNGFEEVQSH AFNGTTLISL ELKENIYLEK MHSGAFQGAT GPSILDISST KLQALPSHGL ESIQTLIALS SYSLKTLPSK EKFTSLLVAT LTYPSHCCAF RNLPKKEQNF SFSIFENFSK QCESTVRKAD NETLYSAIFE ENELSGWDYD YGFCSPKTLQ CAPEPDAFNP CEDIMG | |
Greater than 90% as determined by SDS-PAGE. |