missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV Matrix Protein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Matrix protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9542-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
RABV Matrix Protein | |
Research Use Only | |
M | |
Recombinant Protein | |
Yeast | |
MNVLRKIVKK CRDEDTQKPS PVSAPPDDDD LWLPPPEYVP LKELTSKKNM RNFCVNGDVK ACSPNGYSFR ILRHILRSFN EIYSGNHRMI GLVKVVVGLA LSGAPVPEGM NWVYKLRRTL IFQWADSRGP LEGEELEYSQ EITWDDDTEF VGLQIRVSAR QCHIQGRIWC INTNSRACQL WSDMSLQTQR SEEDKDSSLL LE | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
RABV | |
His | |
Tris-based buffer, 50% glycerol |