missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit Tissue factor pathway inhibitor Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Tissue factor pathway inhibitor, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_001095184.1 |
---|---|
Gene ID (Entrez) | 100009401 |
Name | Tissue factor pathway inhibitor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035750
|
enQuireBio™
QP6775-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16055750
|
enQuireBio™
QP6775-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
16045750
|
enQuireBio™
QP6775-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065750
|
enQuireBio™
QP6775-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16075750
|
enQuireBio™
QP6775-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16085750
|
enQuireBio™
QP6775-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001095184.1 | |
Tissue factor pathway inhibitor Protein | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |
100009401 | |
Research Use Only | |
TFPI | |
Recombinant Protein | |
E. coli | |
AAEEDEEFTN ITDIKPPLQK PTHSFCAMKV DDGPCRAYIK RFFFNILTHQ CEEFIYGGCE GNENRFESLE ECKEKCARDY PKMTTKLTFQ KGKPDFCFLE EDPGICRGYI TRYFYNNQSK QCERFKYGGC LGNLNNFESL EECKNTCENP TSDFQVDDHR TQLNTVNNTL INQPTKAPRR WAFHGPSWCL PPADRGLCQA NEIRFFYNAI IGKCRPFKYS GCGGNENNFT SKKACITACK KGFIPKSIKG GLIKTKRKKK KQPVKITYVE TFVKKT | |
Greater than 90% as determined by SDS-PAGE. |