missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pseudomonas Thiol:disulfide interchange protein DsbA Protein

A DNA sequence encoding the Pseudomonas syringae pv. tomato (strain DC3000) Thiol:disulfide interchange protein DsbA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7497-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029010

  • 3405.00€ / 1mg
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_790191.1
Pseudomonas Thiol:disulfide interchange protein DsbA Protein
Research Use Only
dsbA
Recombinant Protein
E. coli
AEPIESGKQY VELTSAVPVA VPGKIEVIEL FWYGCPHCYA FEPTINPWVE KLPSDVNFVR IPAMFGGPWD AHGQLFITLD TMGVEHKVHA AVFEAIQKGG KRLTDKNDMA DFVATQGVNK DDFLKTFDSF AVKGKIAQYK ELAKKYEVTG VPTMIVNGKY RFDLGSAGGP EKTLQVADQL IDKERAAAKA AK
Greater than 90% as determined by SDS-PAGE.
1181950
1 mg
Not determined.
Full Length
Pseudomonas
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers