missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig E-Selectin / CD62e / SELE Protein

A DNA sequence encoding the Sus scrofa (Pig) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6661-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16024900

  • 2650.00€ / 500µg
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_999433.1
E-Selectin / CD62e / SELE Protein
Research Use Only
SELE
Recombinant Protein
Yeast
WSYSASTETM TFDDASAYCQ QRYTHLVAIQ NHAEIEYLNS TFNYSASYYW IGIRKINGTW TWIGTKKALT PEATNWAPGE PNNKQSNEDC VEIYIKRDKD SGKWNDERCS KKKLALCYTA ACTPTSCSGH GECIETINSS TCQCYPGFRG LQCEQVVECD ALENPVNGVV TCPQSLPWNT TCAFECKEGF ELIGPEHLQC TSSGSWDGKK PTCKAVTCDT VGHPQNGDVS CNHSSIGEFA YKSTCHFTCA EGFGLQGPAQ IECTAQGQWT QQAPVCKAVK CPAVSQPKNG LVKFTHSPTG EFTYKSSCAF SCEEGFELRG SAQLACTSQG QWTQEVPSCQ VVQCSSLEVP REINMSCSGE PVFGAVCTFA CPEGWMLNGS VALTCGATGH WSGMLPTCEA PAESKIP
Greater than 90% as determined by SDS-PAGE.
397508
500 μg
Not determined.
Extracellular Domain
Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers