missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig C5a / Complement 5a Protein
A DNA sequence encoding the Sus scrofa (Pig) C5a / Complement 5a, was expressed in the hosts and tags indicated.
452.00€ - 3175.00€
Specifications
Name | C5a / Complement 5a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | C5 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16047471
|
enQuireBio™
QP9020-YE-10UG |
10 μg |
452.00€
10µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16037471
|
enQuireBio™
QP9020-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16077471
|
enQuireBio™
QP9020-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16067471
|
enQuireBio™
QP9020-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16087471
|
enQuireBio™
QP9020-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16057471
|
enQuireBio™
QP9020-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
C5a / Complement 5a Protein | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
C5 | |
Recombinant Protein | |
Yeast | |
MLQKKIEEEA AKYKYAMLKK CCYDGAYRND DETCEERAAR IKIGPKCVKA FKDCCYIANQ VRAEQSHKNI QLGR | |
Greater than 90% as determined by SDS-PAGE. |