missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Lysyl endopeptidase Protein

A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Lysyl endopeptidase, was expressed in the hosts and tags indicated. option, ab

Brand:  enQuireBio™ QP8146-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032901

  • 920.10€ / 100µg

Estimated Shipment: 13-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_252864.1
P. aeruginosa Lysyl endopeptidase Protein
100 μg
Not determined.
Full Length
P. aeruginosa
His-SUMO
Tris-based buffer, 50% glycerol
880208
Greater than 80% as determined by SDS-PAGE.
Research Use Only
prpL
Recombinant Protein
E. coli
AGYRDGFGAS GSCEVDAVCA TQSGTRAYDN ATAAVAKMVF TSSADGGSYI CTGTLLNNGN SPKRQLFWSA AHCIEDQATA ATLQTIWFYN TTQCYGDAST INQSVTVLTG GANILHRDAK RDTLLLELKR TPPAGVFYQG WSATPIANGS LGHDIHHPRG DAKKYSQGNV SAVGVTYDGH TALTRVDWPS AVVEGGSSGS GLLTVAGDGS YQLRGGLYGG PSYCGAPTSQ RNDYFSDFSG VYSQISRYFA P
Videos
SDS
Documents
Certificates
Special Offers

Special Offers