missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant P. aeruginosa Beta-lactamase OXA-10 Protein

A DNA sequence encoding the Pseudomonas aeruginosa Beta-lactamase OXA-10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7059-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16027270

Quantity Price
1 1665.05€ / 200µg
3 1566.48€ / 200µg
Estimated Shipment: 14-01-2022

Add to basket


Description

Description

Specifications

Specifications

WP_000846390.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
bla
Recombinant Protein
E. coli
SITENTSWNK EFSAEAVNGV FVLCKSSSKS CATNDLARAS KEYLPASTFK IPNAIIGLET GVIKNEHQVF KWDGKPRAMK QWERDLTLRG AIQVSAVPVF QQIAREVGEV RMQKYLKKFS YGNQNISGGI DKFWLEGQLR ISAVNQVEFL ESLYLNKLSA SKENQLIVKE ALVTEAAPEY LVHSKTGFSG VGTESNPGVA WWVGWVEKET EVYFFAFNMD IDNESKLPLR KSIPTKIMES EGIIGG
P. aeruginosa Beta-lactamase OXA-10 Protein
200 μg
Not determined.
Full Length
P. aeruginosa
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents
Certificates
Special Offers

Special Offers