missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mycoplasma Uncharacterized lipoprotein MPN_083 Protein
A DNA sequence encoding the Mycoplasma pneumoniae (strain ATCC 29342 / M129) Uncharacterized lipoprotein MPN_083, was expressed in the hosts and tags indicated.
440.00€ - 3745.00€
Specifications
Accession Number | NP_109771.1 |
---|---|
Gene ID (Entrez) | 877109 |
Name | Mycoplasma Uncharacterized lipoprotein MPN_083 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016880
|
enQuireBio™
QP6955-10UG |
10 μg |
440.00€
10µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16056880
|
enQuireBio™
QP6955-50UG |
50 μg |
630.00€
50µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16006880
|
enQuireBio™
QP6955-100UG |
100 μg |
1070.00€
100µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16036880
|
enQuireBio™
QP6955-200UG |
200 μg |
1765.00€
200µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16046880
|
enQuireBio™
QP6955-500UG |
500 μg |
2360.00€
500µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
16026880
|
enQuireBio™
QP6955-1MG |
1 mg |
3745.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Specifications
NP_109771.1 | |
Mycoplasma Uncharacterized lipoprotein MPN_083 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mycoplasma | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
877109 | |
Research Use Only | |
MPN_083 | |
Recombinant Protein | |
E. coli | |
CSFKDYIPTP SFRKDFSTEN NFVKNKVPGK DDIYSKFYDL TFSLNFVNNQ AQEFGTGWLI DWKGDENKNL SKNKEGQTAS QTRSSSEQTT DQDANLFTAY IATNLHVADG LKNDQDYAPY NKDGWGQPYP YQQKTQSFLL GKYTKPNVQL VKTNYEKPED AVIEQKLKED SLLFIQTSTL PKTAYAAIDP VNFSYNPTRT NGFWTAGKYN VYNGGNSIGN Y | |
Greater than 90% as determined by SDS-PAGE. |