missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Tryptase Protein

A DNA sequence encoding the Mus musculus (Mouse) Tryptase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6831-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036140

  • 770.00€ / 100µg
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_112464.4
Tryptase Protein
Research Use Only
Tpsab1
Recombinant Protein
Yeast
AHGNKWPWQV SLRANDTYWM HFCGGSLIHP QWVLTAAHCV GPDVADPNKV RVQLRKQYLY YHDHLMTVSQ IITHPDFYIV QDGADIALLK LTNPVNISDY VHPVPLPPAS ETFPSGTLCW VTGWGNIDNG VNLPPPFPLK EVQVPIIENH LCDLKYHKGL ITGDNVHIVR DDMLCAGNEG HDSCQGDSGG PLVCKVEDTW LQAGVVSWGE GCAQPNRPGI YTRVTYYLDW IHH
Greater than 90% as determined by SDS-PAGE.
100503895
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers