missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TPT1 / Tumor Protein
A DNA sequence encoding the Mus musculus (Mouse) TPT1 / Tumor, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_033455.1 |
---|---|
Gene ID (Entrez) | 22070 |
Name | TPT1 / Tumor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16076160
|
enQuireBio™
QP6835-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16096160
|
enQuireBio™
QP6835-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16086160
|
enQuireBio™
QP6835-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16006170
|
enQuireBio™
QP6835-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16016170
|
enQuireBio™
QP6835-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16026170
|
enQuireBio™
QP6835-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_033455.1 | |
TPT1 / Tumor Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
22070 | |
Research Use Only | |
Tpt1 | |
Recombinant Protein | |
E. coli | |
MIIYRDLISH DELFSDIYKI REIADGLCLE VEGKMVSRTE GAIDDSLIGG NASAEGPEGE GTESTVVTGV DIVMNHHLQE TSFTKEAYKK YIKDYMKSLK GKLEEQKPER VKPFMTGAAE QIKHILANFN NYQFFIGENM NPDGMVALLD YREDGVTPFM IFFKDGLEME KC | |
Greater than 90% as determined by SDS-PAGE. |