missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TGFB1 Protein
A cDNA sequence encoding the TGFB1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | TGFB1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15924529
|
enQuireBio™
QP13732-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15904529
|
enQuireBio™
QP13732-10UG |
10 μg |
211.00€
10µg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
15914529
|
enQuireBio™
QP13732-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Specifications
TGFB1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
TGFB1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSALDTNYC FSSTEKNCCV RQLYIDFRKD LGWKWIHEPK GYHANFCLGP CPYIWSLDTQ YSKVLALYNQ HNPGASASPC CVPQALEPLP IVYYVGRKPK VEQLSNMIVR SCKCS | |
Greater than 85.0% as determined by SDS-PAGE. |