missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse SOD1 / Superoxide Dismutase Protein
A DNA sequence encoding the Mus musculus (Mouse) SOD1 / Superoxide Dismutase, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_035564.1 |
---|---|
Gene ID (Entrez) | 20655 |
Name | SOD1 / Superoxide Dismutase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035330
|
enQuireBio™
QP6715-YE-100UG |
100 μg |
765.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16055330
|
enQuireBio™
QP6715-YE-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16045330
|
enQuireBio™
QP6715-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065330
|
enQuireBio™
QP6715-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16075330
|
enQuireBio™
QP6715-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16085330
|
enQuireBio™
QP6715-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_035564.1 | |
SOD1 / Superoxide Dismutase Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20655 | |
Research Use Only | |
sod1 | |
Recombinant Protein | |
Yeast | |
AMKAVCVLKG DGPVQGTIHF EQKASGEPVV LSGQITGLTE GQHGFHVHQY GDNTQGCTSA GPHFNPHSKK HGGPADEERH VGDLGNVTAG KDGVANVSIE DRVISLSGEH SIIGRTMVVH EKQDDLGKGG NEESTKTGNA GSRLACGVIG IAQ | |
Greater than 90% as determined by SDS-PAGE. |