missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse REG3G / PAP1B Protein
A DNA sequence encoding the Mus musculus (Mouse) REG3G / PAP1B, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_035390.1 |
---|---|
Gene ID (Entrez) | 19695 |
Name | REG3G / PAP1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084410
|
enQuireBio™
QP6595-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16024420
|
enQuireBio™
QP6595-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16094410
|
enQuireBio™
QP6595-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16014420
|
enQuireBio™
QP6595-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16034420
|
enQuireBio™
QP6595-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16004420
|
enQuireBio™
QP6595-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_035390.1 | |
REG3G / PAP1B Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
19695 | |
Research Use Only | |
Reg3g | |
Recombinant Protein | |
Yeast | |
EVAKKDAPSS RSSCPKGSRA YGSYCYALFS VSKNWYDADM ACQKRPSGHL VSVLSGAEAS FLSSMIKSSG NSGQYVWIGL HDPTLGYEPN RGGWEWSNAD VMNYINWETN PSSSSGNHCG TLSRASGFLK WRENYCNLEL PYVCKFKA | |
Greater than 90% as determined by SDS-PAGE. |