missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein disulfide-isomerase A2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein disulfide-isomerase A2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6479-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001074539.1 | |
Protein disulfide-isomerase A2 Protein | |
Research Use Only | |
Pdia2 | |
Recombinant Protein | |
Yeast | |
QGEEPGGPSE VLPEEPTGEE VPKEDGILVL NHRTLSLALQ EHSALMVEFY APWCGHCKEL APEYSKAAAL LAAESAVVTL AKVDGPAEPE LTKEFEVVGY PTLKFFQNGN RTNPEEYAGP KTAEGIAEWL RRRVGPSATH LEDEEGVQAL MAKWDMVVIG FFQDLQGKDM ATFLALAKDA LDMTFGFTDQ PQLFEKFGLT KDTVVLFKKF DEGRADFPVD KETGLDLGDL SRFLVIHSMH LVTEFNSQTS PKIFAAKILN HLLLFVNQTL AQHRELLTDF REAAPPFRGQ VLFVMVDVAA DNSHVLNYFG LKAEEAPTLR LINVETTKKY APTGVIAITA ASVAAFCQAV LHGEIKHYLL SQEIPPDWDQ GPVKTLVSKN FEQVAFDETK NVFVKFYAPW CSHCKEMAPA WEALAEKYKD REDIVIAELD ATANELEAFS VLGYPTLKFF PAGPDRKVID YKSTRDLETF SKFLDSGGHL PKEEPKEPAA SAPEAQANST LGPKEEL | |
Greater than 90% as determined by SDS-PAGE. |
69191 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |